accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1125-78-6

IUPAC Name: 5,6,7,8-tetrahydronaphthalen-2-ol
Synonyms: tetralol ac-β-tetralol
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: UMKXSOXZAXIOPJ-UHFFFAOYSA-N
SMILES: OC1=CC=C2CCCCC2=C1
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)