accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1125-99-1

IUPAC Name: 1-(cyclohex-1-en-1-yl)pyrrolidine
Synonyms: 1-(cyclohex-1-en-1-yl)pyrrolidine
Molecular Weight (g/mol): 151.25
Molecular Formula: C10H17N
InChi Key: KTZNVZJECQAMBV-UHFFFAOYSA-N
SMILES: C1CCN(C1)C1=CCCCC1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)