accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3(S)NHO(R)NHONHNO(S)HNONNHO(S)HOHNO(R)NNHO(R)ClHNO(R)HNH3COOHN(S)NHH3CCH3ON(S)OHN(R)CH3H2NO

CAS RN 112568-12-4

IUPAC Name: (2S)-N-[(1R)-1-{[(1S)-1-{[(2S)-1-[(2S)-2-{[(1R)-1-carbamoylethyl]carbamoyl}pyrrolidin-1-yl]-1-oxo-6-[(propan-2-yl)amino]hexan-2-yl]carbamoyl}-3-methylbutyl]carbamoyl}-5-[(pyridin-3-yl)formamido]pentyl]-2-[(2S)-2-[(2R)-2-[(2R)-3-(4-chlorophenyl)-2-[(2R)-2-acetamido-3-(naphthalen-2-yl)propanamido]propanamido]-3-(pyridin-3-yl)propanamido]-3-hydroxypropanamido]-6-[(pyridin-3-yl)formamido]hexanamide
Synonyms: (2S)-N-[(1R)-1-{[(1S)-1-{[(2S)-1-[(2S)-2-{[(1R)-1-carbamoylethyl]carbamoyl}pyrrolidin-1-yl]-6-(isopropylamino)-1-oxohexan-2-yl]carbamoyl}-3-methylbutyl]carbamoyl}-5-(pyridin-3-ylformamido)pentyl]-2-[(2S)-2-[(2R)-2-[(2R)-3-(4-chlorophenyl)-2-[(2R)-2-acetamido-3-(naphthalen-2-yl)propanamido]propanamido]-3-(pyridin-3-yl)propanamido]-3-hydroxypropanamido]-6-(pyridin-3-ylformamido)hexanamide
Molecular Weight (g/mol): 1591.32
Molecular Formula: C82H108ClN17O14
InChi Key: QRYFGTULTGLGHU-NBERXCRTSA-N
SMILES: CC(C)C[C@H](NC(=O)[C@@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)[C@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)[C@H](CO)NC(=O)[C@@H](CC1=CC=CN=C1)NC(=O)[C@@H](CC1=CC=C(Cl)C=C1)NC(=O)[C@@H](CC1=CC=C2C=CC=CC2=C1)NC(C)=O)C(=O)N[C@@H](CCCCNC(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@H](C)C(N)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results