accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112582-89-5

IUPAC Name: 6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonamide
Synonyms: 6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonamide
Molecular Weight (g/mol): 237.68
Molecular Formula: C5H4ClN3O2S2
InChi Key: ZVRHRYRTZTWAHL-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=C(Cl)N=C2SC=CN12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)