CAS RN 1126-00-7
IUPAC Name:
1-phenyl-1H-pyrazole
Synonyms:
1-phenyl-1H-pyrazole
Molecular Weight (g/mol):
144.18
Molecular Formula:
C9H8N2
InChi Key:
WITMXBRCQWOZPX-UHFFFAOYSA-N
SMILES:
C1=CN(N=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
1-Phenyl-1H-pyrazole, 98%
CAS: 1126-00-7 Molecular Formula: C9H8N2 Molecular Weight (g/mol): 144.18 MDL Number: MFCD00003112 InChI Key: WITMXBRCQWOZPX-UHFFFAOYSA-N Synonym: 1-phenyl-1h-pyrazole,n-phenylpyrazole,1h-pyrazole, 1-phenyl,pyrazole, 1-phenyl,1-phenylpyrazol,n-phenyl pyrazole,1-phenyl pyrazole,1-phenyl-pyrazole,1-phenyl-1h-pyrazol,l-phenyl-1h-pyrazole PubChem CID: 70769 ChEBI: CHEBI:38879 IUPAC Name: 1-phenylpyrazole SMILES: C1=CN(N=C1)C1=CC=CC=C1