accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHN

CAS RN 1126-78-9

IUPAC Name: N-butylaniline
Synonyms: N-butylaniline
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: VSHTWPWTCXQLQN-UHFFFAOYSA-N
SMILES: CCCCNC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results