accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHSH

CAS RN 1126-81-4

IUPAC Name: N-(4-sulfanylphenyl)acetamide
Synonyms: N-(4-sulfanylphenyl)acetamide
Molecular Weight (g/mol): 167.23
Molecular Formula: C8H9NOS
InChi Key: AYEQJLOHMLYKAV-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(S)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results