CAS RN 112648-68-7
IUPAC Name:
1-(6-{[(1S,3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]amino}hexyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms:
1-(6-{[(1S,3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H-cyclopenta[a]phenanthren-1-yl]amino}hexyl)pyrrole-2,5-dione
Molecular Weight (g/mol):
464.65
Molecular Formula:
C29H40N2O3
InChi Key:
LUFAORPFSVMJIW-ZRJUGLEFSA-N
SMILES:
COC1=CC=C2[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CCC2=C1)NCCCCCCN1C(=O)C=CC1=O
1
–
2
of
2
results
1
Thermo Scientific Chemicals U-73122, 95%
CAS: 112648-68-7 Molecular Formula: C29H40N2O3 Synonym: 1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione
2