accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1127-74-8

IUPAC Name: 2,3,4,5-tetrahydro-1H-1-benzazepin-5-one
Synonyms: 1,2,3,4-tetrahydro-1-benzazepin-5-one
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: NKRKBYFBKLDCFB-UHFFFAOYSA-N
SMILES: O=C1CCCNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results