accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112811-59-3

IUPAC Name: 1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Synonyms: gatifloxacin gatiquin gaity gatifloxacine gatifloxacin, (+-)- gatispan gatifloxacin, (+-)-isomer gatifloxacin (product) tequin gatilox
Molecular Weight (g/mol): 375.4
Molecular Formula: C19H22FN3O4
InChi Key: XUBOMFCQGDBHNK-UHFFFAOYNA-N
SMILES: COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results