accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOFF(E)

CAS RN 112898-33-6

IUPAC Name: (2E)-3-(2,5-difluorophenyl)prop-2-enoate
Synonyms: (2E)-3-(2,5-difluorophenyl)prop-2-enoate
Molecular Weight (g/mol): 183.13
Molecular Formula: C9H5F2O2
InChi Key: XAWHCSKPALFWBI-DAFODLJHSA-M
SMILES: [O-]C(=O)\C=C\C1=CC(F)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results