accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112901-68-5

H2OH2OH2OH2NHNSPOHOHO

CAS RN 112901-68-5

IUPAC Name: ({2-[(3-aminopropyl)amino]ethyl}sulfanyl)phosphonic acid trihydrate
Synonyms: amifostine trihydrate
Molecular Weight (g/mol): 268.26
Molecular Formula: C5H21N2O6PS
InChi Key: TXQPXJKRNHJWAX-UHFFFAOYSA-N
SMILES: O.O.O.NCCCNCCSP(O)(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)