accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OSiO

CAS RN 112926-00-8

IUPAC Name: silanedione
Synonyms: silica 二氧化硅 siliziumdioxid solum diatomeae quartz (SiO2)n kieselsaeureanhydrid SiO2 silica gel silica dimethyl silylate
Molecular Weight (g/mol): 60.08
Molecular Formula: O2Si
InChi Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES: O=[Si]=O
Material 
  • (15)
Quantity 
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (15)
  • (1)
  • (15)
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (10)