accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1130-32-1

IUPAC Name: 3-azaspiro[5.5]undecane-2,4-dione
Synonyms: 3-azaspiro[5.5]undecane-2,4-dione
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: FNIPRNMPSXNBDI-UHFFFAOYSA-N
SMILES: O=C1CC2(CCCCC2)CC(=O)N1
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (2)
  • (5)
  • (8)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (5)
  • (3)