accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1132-21-4

IUPAC Name: 3,5-dimethoxybenzoic acid
Synonyms: 3,5-dimethoxybenzoic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: IWPZKOJSYQZABD-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1)C(O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results