CAS RN 1132-61-2
MOPS (Fine White Crystals/Molecular Biology), Fisher BioReagents™
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N Synonym: 3-(4-Morpholino)propane sulfonic acid PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
MOPS, 99%
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N PubChem CID: 70807 ChEBI: CHEBI:44115 SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
Thermo Scientific Chemicals MOPS, 1.0M buffer soln., pH 7.0
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
Thermo Scientific Chemicals MOPS, 1.0M buffer soln., pH 7.5
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
Thermo Scientific Chemicals MOPS, 99.5%, for molecular biology, Dnase, Rnase, Prot. free, for peptide sequencing
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N Synonym: 3-(N-Morpholino)propanesulfonic acid PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
Thermo Scientific Chemicals MOPS, 0.5M buffer soln., pH 7.0
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
Thermo Scientific Chemicals MOPS, 99%, for biochemistry
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N Synonym: 3-(N-Morpholino)propanesulfonic acid PubChem CID: 70807 ChEBI: CHEBI:44115 SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
Thermo Scientific Chemicals MOPS, 1.0M buffer soln., pH 6.5
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
MOPS, ≥98.5% (Dry Basis), Ultrapure
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N Synonym: 3-[N-Morpholino]propane sulfonic Acid PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: C1COCCN1CCCS(=O)(=O)O
MOPS, 1.0M buffer soln., pH 9.0, Thermo Scientific™
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
MOPS, 1.0M buffer soln., pH 5.5, Thermo Scientific™
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
Thermo Scientific Chemicals MOPS, 0.5M buffer soln., pH 7.5
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
Thermo Scientific Chemicals MOPS, 1.0M buffer soln., pH 8.0
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N PubChem CID: 70807 ChEBI: CHEBI:44115 IUPAC Name: 3-morpholin-4-ylpropane-1-sulfonic acid SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
MOPS, MP Biomedicals™
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N Synonym: 3-(N-morpholino)propanesulfonic acid,4-morpholinepropanesulfonic acid PubChem CID: 70807 ChEBI: CHEBI:44115 SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
MOPS, free acid, 100.2%, MP Biomedicals™
CAS: 1132-61-2 Molecular Formula: C7H15NO4S Molecular Weight (g/mol): 209.26 MDL Number: MFCD00006183 InChI Key: DVLFYONBTKHTER-UHFFFAOYSA-N Synonym: 3-[N-Morpholino]propanesulfonic acid PubChem CID: 70807 ChEBI: CHEBI:44115 SMILES: [O-]S(=O)(=O)CCC[NH+]1CCOCC1
- 1
- 2