accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 113212-14-9

IUPAC Name: methyl (2Z)-3-(dimethylamino)-2-isocyanoprop-2-enoate
Synonyms: methyl (2Z)-3-(dimethylamino)-2-isocyanoprop-2-enoate
Molecular Weight (g/mol): 154.17
Molecular Formula: C7H10N2O2
InChi Key: FREGRQYTSPAJDX-WAYWQWQTSA-N
SMILES: COC(=O)C(=C\N(C)C)\[N+]#[C-]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results