accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOHNH2

CAS RN 113293-71-3

IUPAC Name: (6-aminopyridin-3-yl)methanol
Synonyms: (6-aminopyridin-3-yl)methanol
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: TXPRFSOGPYITOT-UHFFFAOYSA-N
SMILES: NC1=NC=C(CO)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results