accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Br

CAS RN 1133-80-8

IUPAC Name: 2-bromo-9H-fluorene
Synonyms: 2-bromo-9H-fluorene
Molecular Weight (g/mol): 245.12
Molecular Formula: C13H9Br
InChi Key: FXSCJZNMWILAJO-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(CC3=CC=CC=C23)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)