accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNOO

CAS RN 113439-83-1

IUPAC Name: 2-(but-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(but-2-yn-1-yl)isoindole-1,3-dione
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: COPWPECHFWIYSP-UHFFFAOYSA-N
SMILES: CC#CCN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results