accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHOHO(E)

CAS RN 1135-24-6

IUPAC Name: (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
Synonyms: ferulic acid (e)-ferulic acid ferulic acid, (e)-isomer ferulic acid, (E)- ferulic acid, (z)-isomer coniferic acid ferulic acid, (E)-isomer ferulic acid, (z)- (z)-isomer ferulic acid ferulic acid, (e)-
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: KSEBMYQBYZTDHS-HWKANZROSA-N
SMILES: COC1=CC(\C=C\C(O)=O)=CC=C1O
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)