accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClHSnHCl

CAS RN 1135-99-5

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 345.84
Molecular Formula: C12H12Cl2Sn
InChi Key: WSXPROXKZIMZIU-UHFFFAOYSA-N
SMILES: Cl.Cl.[SnH2](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)