CAS RN 113512-57-5
IUPAC Name:
2,4-difluoro-5-nitrophenol
Synonyms:
2,4-difluoro-5-nitrophenol
Molecular Weight (g/mol):
175.09
Molecular Formula:
C6H3F2NO3
InChi Key:
SMRYCTJAGPDVEH-UHFFFAOYSA-N
SMILES:
OC1=C(F)C=C(F)C(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
2,4-Difluoro-5-nitrophenol, 98%
CAS: 113512-57-5 Molecular Formula: C6H3F2NO3 Molecular Weight (g/mol): 175.091 MDL Number: MFCD08443942 InChI Key: SMRYCTJAGPDVEH-UHFFFAOYSA-N Synonym: 2,4-difluoro-5-nitrobenzenol,phenol, 2,4-difluoro-5-nitro,difluoronitrobenzenol,acmc-209wyr,2,4-difluoro-5-hydroxynitrobenzene PubChem CID: 10921024 IUPAC Name: 2,4-difluoro-5-nitrophenol SMILES: C1=C(C(=CC(=C1O)F)F)[N+](=O)[O-]