accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHNNNOOCH3CH3CH3HCl

CAS RN 1135210-68-2

IUPAC Name: ethyl 4-(butylamino)-1-ethyl-6-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate hydrochloride
Synonyms: hydrochloride tracazolate
Molecular Weight (g/mol): 340.85
Molecular Formula: C16H25ClN4O2
InChi Key: TUIXDPUEMQXVHO-UHFFFAOYSA-N
SMILES: Cl.CCCCNC1=C2C=NN(CC)C2=NC(C)=C1C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results