accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNNN

CAS RN 1137-67-3

IUPAC Name: 2-(pyridin-3-yl)-1H-1,3-benzodiazole
Synonyms: 2-(pyridin-3-yl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 195.23
Molecular Formula: C12H9N3
InChi Key: BOUOQESVDURNSB-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2N=C1C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results