accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1138-80-3

HOONHOO

CAS RN 1138-80-3

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}acetic acid
Synonyms: (cbz)gly carbobenzoxyl glycine Z-gly N-carbobenzyloxyglycine carbobenzyloxyglycine N-cbz-glycine
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: CJUMAFVKTCBCJK-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)