accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHNHHNHNCH3HClHClHClHCl

CAS RN 113812-15-0

IUPAC Name: 3,7,12,16-tetraazaoctadecane tetrahydrochloride
Synonyms: 3,7,12,16-tetraazaoctadecane tetrahydrochloride
Molecular Weight (g/mol): 404.29
Molecular Formula: C14H38Cl4N4
InChi Key: SCYOBNRPMHJGLB-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.CCNCCCNCCCCNCCCNCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results