CAS RN 113841-59-1
IUPAC Name:
4-(bromomethyl)-3-methyl-5-phenyl-1,2-oxazole
Synonyms:
4-(bromomethyl)-3-methyl-5-phenyl-1,2-oxazole
Molecular Weight (g/mol):
252.11
Molecular Formula:
C11H10BrNO
InChi Key:
KCGVZUOBUPWFJC-UHFFFAOYSA-N
SMILES:
CC1=NOC(=C1CBr)C1=CC=CC=C1
1
–
1
of
1
results
1
4-(Bromomethyl)-3-methyl-5-phenylisoxazole, ≥95%, Thermo Scientific™
CAS: 113841-59-1 Molecular Formula: C11H10BrNO Molecular Weight (g/mol): 252.111 MDL Number: MFCD03086124 InChI Key: KCGVZUOBUPWFJC-UHFFFAOYSA-N PubChem CID: 2776518 IUPAC Name: 4-(bromomethyl)-3-methyl-5-phenyl-1,2-oxazole SMILES: CC1=NOC(=C1CBr)C2=CC=CC=C2