accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 113975-33-0

CH3H3CCH3ONHNI

CAS RN 113975-33-0

IUPAC Name: N-(3-iodopyridin-4-yl)-2,2-dimethylpropanamide
Synonyms: N-(3-iodopyridin-4-yl)-2,2-dimethylpropanamide
Molecular Weight (g/mol): 304.13
Molecular Formula: C10H13IN2O
InChi Key: GPMKCDBJLNTANL-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)NC1=CC=NC=C1I
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results