accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 114-49-8

BrH3CN(S)(S)(S)O(R)OO(S)OH

CAS RN 114-49-8

IUPAC Name: (1S,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-7-yl (2S)-3-hydroxy-2-phenylpropanoate hydrobromidyl
Synonyms: (1S,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-7-yl (2S)-3-hydroxy-2-phenylpropanoate hydrobromidyl
Molecular Weight (g/mol): 383.26
Molecular Formula: C17H21BrNO4
InChi Key: XUZPYCBMTSSDHK-AKTDCHNFSA-N
SMILES: [Br].CN1[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)[C@H](CO)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results