accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 114-63-6

IUPAC Name: sodium 4-hydroxybenzoate
Synonyms: sodium methylparaben
Molecular Weight (g/mol): 160.1
Molecular Formula: C7H5NaO3
InChi Key: ZLVSYODPTJZFMK-UHFFFAOYSA-M
SMILES: [Na+].OC1=CC=C(C=C1)C([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results