accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 114-76-1

IUPAC Name: sodium 2-oxo-3-phenylpropanoate
Synonyms: sodium β-phenylpyruvate
Molecular Weight (g/mol): 186.14
Molecular Formula: C9H7NaO3
InChi Key: MQGYVGKMCRDEAF-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C(=O)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results