CAS RN 1141-38-4
IUPAC Name:
naphthalene-2,6-dicarboxylic acid
Synonyms:
2,6-dicarboxynaphthalene 2,6-naphthalic acid
Molecular Weight (g/mol):
216.19
Molecular Formula:
C12H8O4
InChi Key:
RXOHFPCZGPKIRD-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC2=CC=C(C=C2C=C1)C(O)=O
1
–
1
of
1
results
1
Naphthalene-2,6-dicarboxylic acid, 98+%
CAS: 1141-38-4 Molecular Formula: C12H8O4 Molecular Weight (g/mol): 216.192 MDL Number: MFCD00004105 InChI Key: RXOHFPCZGPKIRD-UHFFFAOYSA-N Synonym: 2,6-naphthalenedicarboxylic acid,2,6-naphthalic acid,2,6-dicarboxynaphthalene,unii-k3c4dyz29o,2,6-naphthalene dicarboxylic acid,k3c4dyz29o,dsstox_cid_9211,acmc-2099kj,d06kmp,dsstox_rid_78711 PubChem CID: 14357 ChEBI: CHEBI:44460 IUPAC Name: naphthalene-2,6-dicarboxylic acid SMILES: C1=CC2=C(C=CC(=C2)C(=O)O)C=C1C(=O)O