accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1141-88-4

IUPAC Name: 2-[(2-aminophenyl)disulfanyl]aniline
Synonyms: benzenamine, 2,2'-dithiobis-
Molecular Weight (g/mol): 248.36
Molecular Formula: C12H12N2S2
InChi Key: YYYOQURZQWIILK-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1SSC1=CC=CC=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results