accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 114322-14-4

IUPAC Name: 2,1,3-benzoxadiazole-4-sulfonyl chloride
Synonyms: 2,1,3-benzoxadiazole-4-sulfonyl chloride
Molecular Weight (g/mol): 218.61
Molecular Formula: C6H3ClN2O3S
InChi Key: SIYWVPMYOGMRDW-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=CC2=NON=C12
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)