accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CCH3OONHOHONHOO

CAS RN 114360-56-4

IUPAC Name: 4-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: 4-[(tert-butoxycarbonyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 440.5
Molecular Formula: C24H28N2O6
InChi Key: LIWKOFAHRLBNMG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)