CAS RN 1144-74-7
IUPAC Name:
(4-nitrophenyl)(phenyl)methanone
Synonyms:
(4-nitrophenyl)(phenyl)methanone
Molecular Weight (g/mol):
227.22
Molecular Formula:
C13H9NO3
InChi Key:
ZYMCBJWUWHHVRX-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Nitrobenzophenone, 99%
CAS: 1144-74-7 Molecular Formula: C13H9NO3 Molecular Weight (g/mol): 227.22 MDL Number: MFCD00007354 InChI Key: ZYMCBJWUWHHVRX-UHFFFAOYSA-N Synonym: 4-nitrobenzophenone,p-nitrobenzophenone,4-nitrophenyl phenyl methanone,benzophenone, 4-nitro,methanone, 4-nitrophenyl phenyl,4-nitrophenyl phenyl ketone,4-nitrophenyl-phenylmethanone,4-nitrophenyl-phenyl-methanone,4-nitro-benzophenone PubChem CID: 70839 IUPAC Name: (4-nitrophenyl)-phenylmethanone SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1