accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1145-98-8

CH3SiH3CSiCH3H3C

CAS RN 1145-98-8

IUPAC Name: 1,1,2,2-tetramethyl-1,2-diphenyldisilane
Synonyms: 1,1,2,2-tetramethyl-1,2-diphenyldisilane
Molecular Weight (g/mol): 270.52
Molecular Formula: C16H22Si2
InChi Key: IIOOIYHUTINYQO-UHFFFAOYSA-N
SMILES: C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results