accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOSOO

CAS RN 1146-39-0

IUPAC Name: 1-(benzenesulfonyl)-4-nitrobenzene
Synonyms: 1-(benzenesulfonyl)-4-nitrobenzene
Molecular Weight (g/mol): 263.27
Molecular Formula: C12H9NO4S
InChi Key: OECHJYYZMSUILG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results