accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNH2

CAS RN 114715-38-7

IUPAC Name: 1-benzylpyrrolidin-3-amine
Synonyms: 1-benzylpyrrolidin-3-amine
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: HBVNLKQGRZPGRP-UHFFFAOYNA-N
SMILES: NC1CCN(CC2=CC=CC=C2)C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results