accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 114776-15-7

IUPAC Name: 2-chloro-4-fluoro-5-nitrobenzoate
Synonyms: 2-chloro-4-fluoro-5-nitrobenzoate
Molecular Weight (g/mol): 218.54
Molecular Formula: C7H2ClFNO4
InChi Key: SYZKAFCPWNFONG-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=C(F)C=C1Cl)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results