accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 114897-92-6

BrFOHO

CAS RN 114897-92-6

IUPAC Name: 2-(4-bromo-2-fluorophenyl)acetic acid
Synonyms: (4-bromo-2-fluorophenyl)acetic acid
Molecular Weight (g/mol): 233.04
Molecular Formula: C8H6BrFO2
InChi Key: PNBIYFPZODYMOO-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(F)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results