accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONH2FFF

CAS RN 114926-38-4

IUPAC Name: 2-amino-3-[4-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 2-amino-3-[4-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 233.19
Molecular Formula: C10H10F3NO2
InChi Key: CRFFPDBJLGAGQL-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(C=C1)C(F)(F)F)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)