accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 115-21-9

H3CSiClClCl

CAS RN 115-21-9

IUPAC Name: trichloro(ethyl)silane
Synonyms: ethyltrichlorosilane
Molecular Weight (g/mol): 163.5
Molecular Formula: C2H5Cl3Si
InChi Key: ZOYFEXPFPVDYIS-UHFFFAOYSA-N
SMILES: CC[Si](Cl)(Cl)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results