accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OSOOCl

CAS RN 115010-11-2

IUPAC Name: 2,3-dihydro-1-benzofuran-5-sulfonyl chloride
Synonyms: 2,3-dihydro-1-benzofuran-5-sulfonyl chloride
Molecular Weight (g/mol): 218.65
Molecular Formula: C8H7ClO3S
InChi Key: RVWYPBARHGPULM-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC2=C(OCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)