CAS RN 115066-14-3
6-Cyano-7-nitro-1,4-dihydroquinoxaline-2,3-dione, 98%
CAS: 115066-14-3 Molecular Formula: C9H4N4O4 Molecular Weight (g/mol): 232.155 MDL Number: MFCD00069232 InChI Key: RPXVIAFEQBNEAX-UHFFFAOYSA-N Synonym: cnqx,6-cyano-7-nitroquinoxaline-2,3-dione,unii-6ote87sccw,7-nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile,6ote87sccw,3h cnqx,6-cyano-2,3-dihydroxy-7-nitroquinoxaline,1,4-dihydro-2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile,6-quinoxalinecarbonitrile, 1,2,3,4-tetrahydro-7-nitro-2,3-dioxo,2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile PubChem CID: 3721046 ChEBI: CHEBI:34468 IUPAC Name: 7-nitro-2,3-dioxo-1,4-dihydroquinoxaline-6-carbonitrile SMILES: C1=C(C(=CC2=C1NC(=O)C(=O)N2)[N+](=O)[O-])C#N
R&D Systems™ CNQX
CAS: 115066-14-3 Molecular Formula: C9H4N4O4
6-Cyano-7-nitroquinoxaline-2,3-dione, TRC
CAS: 115066-14-3 Molecular Formula: C9 H4 N4 O4 Molecular Weight (g/mol): 232.15 IUPAC Name: 7-nitro-2,3-dioxo-1,4-dihydroquinoxaline-6-carbonitrile SMILES: [O-][N+](=O)c1cc2NC(=O)C(=O)Nc2cc1C#N