CAS RN 115250-38-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2R,3S,4S,5S) -5-Hydroxy-2,3,4-tris(phenylmethoxy)-5-[(phenylmethoxy)methyl]-cyclohexanone, TRC
CAS: 115250-38-9 Molecular Formula: C35H36O6 Molecular Weight (g/mol): 552.66 Synonym: [2R-(2alpha,3beta,4alpha,5alpha)]- 5-Hydroxy-2,3,4-tris(phenylmethoxy)-5-[(phenylmethoxy)methyl]-cyclohexanone IUPAC Name: (2R,3S,4S,5S)-5-hydroxy-2,3,4-tris(phenylmethoxy)-5-(phenylmethoxymethyl)cyclohexan-1-one SMILES: O[C@]1(COCc2ccccc2)CC(=O)[C@H](OCc3ccccc3)[C@@H](OCc4ccccc4)[C@@H]1OCc5ccccc5