CAS RN 115262-01-6
IUPAC Name:
(bromodifluoromethyl)trimethylsilane
Synonyms:
(bromodifluoromethyl)trimethylsilane
Molecular Weight (g/mol):
203.1
Molecular Formula:
C4H9BrF2Si
InChi Key:
WDZVWBWAUSUTTO-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C(F)(F)Br
1
–
3
of
3
results
1
Trimethyl(bromodifluoromethyl)silane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Trimethyl(bromodifluoromethyl)silane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Trimethyl(bromodifluoromethyl)silane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.