accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 115754-20-6

OHOClFFF

CAS RN 115754-20-6

IUPAC Name: 3-chloro-4-(trifluoromethyl)benzoic acid
Synonyms: 3-chloro-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: UDXPRKSPAZWHQN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results