accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO

CAS RN 1159-86-0

IUPAC Name: (2-benzoylphenyl)(phenyl)methanone
Synonyms: (2-benzoylphenyl)(phenyl)methanone
Molecular Weight (g/mol): 286.33
Molecular Formula: C20H14O2
InChi Key: OJLABXSUFRIXFL-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=CC=C1C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results